Learn More
5-Chloro-2-hydroxybenzyl alcohol, 98%
Product Code. 11384626
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384626 1 g 1g
11374626 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11384626 Supplier Thermo Scientific Alfa Aesar Supplier No. H32533.03
Request Bulk or Custom Format
CAS: 5330-38-1 | C7H7ClO2 | 158.581 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5330-38-1
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00021967
InChI Key AFPSETMCMFRVPG-UHFFFAOYSA-N
Synonym 5-chlorosalicyl alcohol, 4-chloro-2-hydroxymethyl phenol, 5-chloro-2-hydroxybenzyl alcohol, benzenemethanol, 5-chloro-2-hydroxy, benzenemethanol,5-chloro-2-hydroxy, 5-chlorosaligenin, 5-chlorosalicylalcohol, saligenin, 5-chloro, 5-chloro-2-hydroxyphenyl methanol, 5-chloro-2-hydroxyphenylmethyl carbinol
PubChem CID 79238
IUPAC Name 4-chloro-2-(hydroxymethyl)phenol
SMILES C1=CC(=C(C=C1Cl)CO)O
Specifications
Melting Point 84°C to 87°C
Quantity 1 g
Formula Weight 158.58
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-hydroxybenzyl alcohol