Learn More
5-Chloro-1-pentyne, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 14267-92-6 |
| Molecular Formula | C5H7Cl |
| MDL Number | MFCD00001014 |
| Quantity | 25 g |
| SMILES | C#CCCCCl |
| Molecular Weight (g/mol) | 102.56 |
| Percent Purity | 96% |
| Chemical Name or Material | 5-Chloro-1-pentyne |