missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Carbamoylthiophene-2-boronic acid pinacol ester, 96%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Carbamoylthiophene-2-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
5-Carbamoylthiophene-2-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Specifications
| Quantity | 250 mg |
| Formula Weight | 253.13 |
| Percent Purity | 96% |
| Chemical Name or Material | 5-Carbamoylthiophene-2-boronic acid pinacol ester |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?