Learn More
5-Bromoimidazo[1,2-a]pyridine, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 69214-09-1 |
| Molecular Formula | C7H5BrN2 |
| MDL Number | MFCD08275698 |
| Quantity | 5 g |
| SMILES | BrC1=CC=CC2=NC=CN12 |
| Molecular Weight (g/mol) | 197.03 |
| Percent Purity | 96% |
| Chemical Name or Material | 5-Bromoimidazo[1,2-a]pyridine |