missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolylphosphate Disodium Salt, ≥98%, MP Biomedicals™

Product Code. 11416442 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416442 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416442 Supplier MP Biomedicals Supplier No. 0215004283
  • ONLINE EXCLUSIVE PRICE
    212.00 EUR
    201.60 EUR / 250mg
    Save 10.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disodium salt is soluble in water (50mg/mL); insoluble in dimethylformamide

Specifications

CAS 102185-33-1
Color White
Assay Percent Range ≥98%
Molecular Formula C8H4BrClNNa2O4P
MDL Number MFCD00036757
Quantity 250 mg
Synonym sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip™, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
IUPAC Name disodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
Molecular Weight (g/mol) 370.43
PubChem CID 6097197
Formula Weight 370.4
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolylphosphate Disodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.