missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-β-D-glucuronide Sodium Salt, MP Biomedicals™

Product Code. 11446341 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446341 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11446341 Supplier MP Biomedicals Supplier No. 0219392505
  • ONLINE EXCLUSIVE PRICE
    65.05 EUR
    61.95 EUR / 5mg
    Save 3.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A β-glucuronidase substrate which forms an intense blue precipitate upon enzymatic action. Used for the detection of the GUS gene in bacterial colonies and in histochemical applications

Specifications

CAS 129541-41-9
Color White
Assay Percent Range ≥98%
Molecular Formula C14H12BrClNNaO7
Quantity 5 mg
Synonym x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
InChI Key IBLSVGDGSKUDCT-ILIJQVQCSA-M
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]
IUPAC Name sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol) 444.594
PubChem CID 53384407
Formula Weight 444.6
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.