missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, MP Biomedicals™

Product Code. 11486392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486392 250 mg 250mg
11406402 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486392 Supplier MP Biomedicals Supplier No. 0215000183
  • ONLINE EXCLUSIVE PRICE
    185.00 EUR
    175.35 EUR / 250mg
    Save 9.65 EUR
    5% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly known as X-Gal. 5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, MP Biomedicals is a histochemical substrate for β-galactosidase. It is particularly useful in molecular biology, where β-galactosidase is frequently used as a report

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques.

It is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

  • Insertion of DNA into the lac Z region results in the loss of β-galactosidase activity
  • Lac+ bacterial colonies resulting from a-complementation will appear blue
  • Bacterial colonies containing plasmid with DNA inserted in the lac Z region will be incapable of a-complementation and will appear white

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Melting Point 230°C (decomposition)(literature)
Quantity 250 mg
Solubility Information Soluble in DMF (2% w/v, clear and colorless solution).
Formula Weight 408.6
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.