missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methyl-2-(methylamino)pyridine, 96%, Thermo Scientific™

Product Code. 15430678
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430678 1 g 1g
15440678 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430678 Supplier Thermo Scientific Alfa Aesar Supplier No. H54360.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 245765-66-6
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.067
MDL Number MFCD10000482
InChI Key QTBNIBCNVQMGPP-UHFFFAOYSA-N
Synonym 5-bromo-3-methyl-2-methylamino pyridine, 5-bromo-n,3-dimethyl-2-pyridinamine, 2-pyridinamine, 5-bromo-n,3-dimethyl, 5-bromo-3-methyl-pyridin-2-yl-methyl-amine
PubChem CID 18329367
IUPAC Name 5-bromo-N,3-dimethylpyridin-2-amine
SMILES CC1=CC(=CN=C1NC)Br

Specifications

Melting Point 62°C to 64°C
Quantity 1 g
Formula Weight 201.06
Percent Purity 96%
Chemical Name or Material 5-Bromo-3-methyl-2-(methylamino)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.