accessibility menu, dialog, popup

UserName

CAS RN 245765-66-6

CH3NHNCH3Br

CAS RN 245765-66-6

IUPAC Name: 5-bromo-N,3-dimethylpyridin-2-amine
Synonyms: 5-bromo-N,3-dimethylpyridin-2-amine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: QTBNIBCNVQMGPP-UHFFFAOYSA-N
SMILES: CNC1=NC=C(Br)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results