missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-phenylpyridine, 95%, Thermo Scientific™

Product Code. 15453787
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453787 250 mg 250mg
11397379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453787 Supplier Thermo Scientific Alfa Aesar Supplier No. H27816.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-phenylpyridine is used as a ligand in coordination chemistry and reacts with iridium(III) chloride to get in tris-heteroleptic Iridium(III), which finds application as phosphorescent (triplet) emitters in organic light-emitting diodes (OLEDs) due to its self-emitting nature, fast response, wide viewing angle and low power consumption.

Solubility
Soluble in ethyl ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27012-25-5
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD04114181
InChI Key PRNGIODVYLTUKH-UHFFFAOYSA-N
Synonym 2-phenyl-5-bromopyridine, 3-bromo-6-phenylpyridine, pyridine, 5-bromo-2-phenyl, 5-bromo-2-phenyl-pyridine, pubchem21997, acmc-20am5l, 5-bromo-2-phenylpyridine, abbypharma ap-11-5994
PubChem CID 2762862
IUPAC Name 5-bromo-2-phenylpyridine
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)Br

Specifications

Melting Point 73°C to 75°C
Quantity 250 mg
Solubility Information Soluble in ethyl ether.
Formula Weight 234.1
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.