Learn More
5-Bromo-2-fluorobenzylamine hydrochloride, 97%
Product Code. 15463277
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463277 1 g 1g
11460628 5 g 5g
11470628 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15463277 Supplier Thermo Scientific Alfa Aesar Supplier No. B25176.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 202865-69-8 | C7H8BrFN | 205.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202865-69-8
Molecular Formula C7H8BrFN
Molecular Weight (g/mol) 205.05
MDL Number MFCD00143426
InChI Key PDKBJZGGXHTHNC-UHFFFAOYSA-O
Synonym 5-bromo-2-fluorobenzylamine hydrochloride, 5-bromo-2-fluorophenyl methanamine hydrochloride, 5-bromo-2-fluorobenzylamine hcl, 5-bromo-2-fluorophenyl methanamine hcl, 1-5-bromo-2-fluorophenyl methanamine hydrochloride, pubchem3227, acmc-1cpb5, rarechem al bd 0325, akos bbv-005136, 5-bromo-2-fluorobenzyl amine hcl
PubChem CID 2724903
SMILES [NH3+]CC1=CC(Br)=CC=C1F
Specifications
Melting Point 221°C to 226°C
Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name (5-bromo-2-fluorophenyl)methanamine;hydrochloride
Formula Weight 240.5
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluorobenzylamine hydrochloride