missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydroxybenzoic Acid, TRC

Product Code. 30343732
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343732 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343732 Supplier Toronto Research Chemicals Supplier No. TRCB68412510G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromo-2,3-dihydroxybenzoic Acid
CAS 72517-15-8
Molecular Formula C7 H5 Br O4
Quantity 10 g
Synonym 5-Bromo-o-pyrocatechuic Acid
InChI Formula InChI=1S/C7H5BrO4/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,9-10H,(H,11,12)
SMILES OC(=O)c1cc(Br)cc(O)c1O
IUPAC Name 5-bromo-2,3-dihydroxybenzoic acid
Molecular Weight (g/mol) 233.02
Formula Weight 231.9371
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.