Learn More
5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Sodium Salt, Mikromol
Product Code. 30463192
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463192 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463192 Supplier Mikromol Supplier No. MM0154.210025
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Sodium Salt, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Sodium Salt
CAS 2469736-46-5
Analyte or Component Names 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Sodium Salt
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H22 N3 O4 . Na
Certifications/Compliance ISO 9001
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-hydroxyethyl)amino]-1-methyl-, sodium salt (1:1) (ACI)
InChI Formula InChI=1S/C16H23N3O4.Na/c1-18-14-6-5-12(19(7-9-20)8-10-21)11-13(14)17-15(18)3-2-4-16(22)23;/h5-6,11,20-21H,2-4,7-10H2,1H3,(H,22,23);/q;+1/p-1
Solution Type Neat
SMILES [Na+].Cn1c(CCCC(=O)[O-])nc2cc(ccc12)N(CCO)CCO
IUPAC Name sodium;4-[5-[bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
Molecular Weight (g/mol) 343.3534
Quantity 25 mg
Formula Weight 343.1508 g/mol