Learn More
5-Benzamidolevulinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Benzamidolevulinic Acid |
| CAS | 102872-00-4 |
| Molecular Formula | C12H13NO4 |
| Quantity | 25 mg |
| Synonym | 5-Benzamido-4-oxopentanoic acid |
| InChI Formula | InChI=1S/C12H13NO4/c14-10(6-7-11(15)16)8-13-12(17)9-4-2-1-3-5-9/h1-5H,6-8H2,(H,13,17)(H,15,16) |
| SMILES | O=C(CCC(O)=O)CNC(C1=CC=CC=C1)=O |
| IUPAC Name | 5-benzamido-4-oxopentanoic acid |
| Molecular Weight (g/mol) | 235.25 |
| Formula Weight | 235.08 |