Learn More
5-Aminoisophthalic Acid, Sigma-Aldrich™
Product Code. 15878500
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878500 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15878500 Supplier Sigma Aldrich Supplier No. 8012460050
 Restricted Item
Estimated Shipment: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 99-31-0
Melting Point 150°C to 156°C
MDL Number MFCD00002522
Quantity 50 g
Synonym 5-Aminoisophthalic acid, 5-Aminoisophthalic acid
Molecular Weight (g/mol) 181.15
Grade Research