Learn More
5-Allylidene-10,11-dihydro-5H-dibenzo[a,d][7]annulene (Stabilized with Hydroquinone), TRC
Specifications
Specifications
| Chemical Name or Material | 5-Allylidene-10,11-dihydro-5H-dibenzo[a,d][7]annulene |
| CAS | 24755-73-5 |
| Molecular Formula | C18 H16 |
| Quantity | 100 mg |
| Synonym | 10,11-Dihydro-5-(2-propenylidene)-5H-dibenzo[a,d]cycloheptene, 5-Allylidene-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, 10,11-Dihydro-5-(2-propen-1-ylidene)-5H-dibenzo[a,d]cycloheptene |
| InChI Formula | InChI=1S/C18H16/c1-2-7-18-16-10-5-3-8-14(16)12-13-15-9-4-6-11-17(15)18/h2-11H,1,12-13H2 |
| SMILES | C=CC=C1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 11-prop-2-enylidene-5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulene |
| Molecular Weight (g/mol) | 232.32 |
| Formula Weight | 232.1252 |