Learn More
5-Acetoxymethyl-2-furancarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Acetoxymethyl-2-furancarboxylic Acid |
| CAS | 90345-66-7 |
| Molecular Formula | C8 H8 O5 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H8O5/c1-5(9)12-4-6-2-3-7(13-6)8(10)11/h2-3H,4H2,1H3,(H,10,11) |
| SMILES | CC(=O)OCc1oc(cc1)C(=O)O |
| IUPAC Name | 5-(acetyloxymethyl)furan-2-carboxylic acid |
| Molecular Weight (g/mol) | 184.15 |
| Formula Weight | 184.0372 |