missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dibromo-8-hydroxyquinoline, MP Biomedicals

Product Code. 11487585 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487585 100 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487585 Supplier MP Biomedicals Supplier No. 0521198525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 521-74-4
Molecular Formula C9H5Br2NO
Quantity 100 g
Synonym broxyquinoline, 5,7-dibromo-8-hydroxyquinoline, 5,7-dibromo-8-quinolinol, broxyquinolin, dibromoxin, dibromoxine, dibromoxyquinoline, broxiquinoline, dibromoquin, broxykinolin
InChI Key ZDASUJMDVPTNTF-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C(C=C2Br)Br)O)N=C1
IUPAC Name 5,7-dibromoquinolin-8-ol
Molecular Weight (g/mol) 302.953
PubChem CID 2453
Formula Weight 302.96
Chemical Name or Material 5, 7-Dibromo-8-hydroxyquinoline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.