Learn More
5,6-Dichloro-1,3-benzoxazol-2-amine, TRC
Product Code. 30376062
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376062 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30376062 Supplier Toronto Research Chemicals Supplier No. TRCD434705100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 64037-12-3
Molecular Formula C7H4Cl2N2O
Quantity 100 mg
InChI Formula InChI=1S/C7H4Cl2N2O/c8-3-1-5-6(2-4(3)9)12-7(10)11-5/h1-2H,(H2,10,11)
SMILES Nc1oc2cc(Cl)c(Cl)cc2n1
Molecular Weight (g/mol) 203.03
Formula Weight 201.9701
Chemical Name or Material 5,6-Dichloro-1,3-benzoxazol-2-amine