Learn More
5,5-Dimethyl-2-isopropyl-1,3-dioxane, TRC
Product Code. 30365498
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365498 500 mg 500mg
30355711 1 g 1g
30351257 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30365498 Supplier Toronto Research Chemicals Supplier No. TRCD356890500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7651-50-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
Synonym 5,5-Dimethyl-2-(1-methylethyl)-1,3-dioxane, 2-Isopropyl-5,5-dimethyl-1,3-dioxane
IUPAC Name 5,5-dimethyl-2-propan-2-yl-1,3-dioxane
SMILES CC(C)C1OCC(C)(C)CO1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C9H18O2/c1-7(2)8-10-5-9(3,4)6-11-8/h7-8H,5-6H2,1-4H3
Formula Weight 158.1307
Chemical Name or Material 5,5-Dimethyl-2-isopropyl-1,3-dioxane