Learn More
5,5-Dimethyl-1,3-dioxane, TRC
Product Code. 30383650
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383650 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30383650 Supplier Toronto Research Chemicals Supplier No. TRCD890803250MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,5-Dimethyl-1,3-dioxane
CAS 872-98-0
Molecular Formula C6 H12 O2
Quantity 250 mg
Synonym 5,5-Dimethyl-m-dioxane, NSC 139437
InChI Formula InChI=1S/C6H12O2/c1-6(2)3-7-5-8-4-6/h3-5H2,1-2H3
SMILES CC1(C)COCOC1
IUPAC Name 5,5-dimethyl-1,3-dioxane
Molecular Weight (g/mol) 116.158
Formula Weight 116.0837