Learn More
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
Product Code. 10374322
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10374322 1 g 1g
10590552 250 mg 250mg
10727285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10374322 Supplier Thermo Scientific Maybridge Supplier No. CC18203DA
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Chemical Identifiers
CAS 215434-25-6
Molecular Formula C8H6ClNO2S3
Molecular Weight (g/mol) 279.771
InChI Key YIKNRCVJBOLZFL-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone
PubChem CID 2736979
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
SMILES CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl
Specifications
Assay Percent Range 95%
Quantity 1 g
Formula Weight 279.79
Percent Purity 95%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride