missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-(2-Bromophenyl)-1H-tetrazole, 98+%

Product Code. 11465077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465077 1 g 1g
11475077 5 g 5g
11485077 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465077 Supplier Alfa Aesar™ Supplier No. B21966.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 73096-42-1
Molecular Formula C7H5BrN4
Molecular Weight (g/mol) 225.049
MDL Number MFCD00151799
InChI Key YHVBXKTXLJTDRI-UHFFFAOYSA-N
PubChem CID 560689
IUPAC Name 5-(2-bromophenyl)-2H-tetrazole
SMILES C1=CC=C(C(=C1)C2=NNN=N2)Br

Specifications

Melting Point 181°C to 183°C
Quantity 1 g
Beilstein 148567
Synonym 5-2-bromophenyl-1h-tetrazole, 5-2-bromophenyl tetrazole, 5-2-bromo-phenyl-2h-tetrazole, 5-2-bromophenyl-2h-tetrazole, 5-2-bromophenyl-1h-1,2,3,4-tetrazole, 5-2-bromophenyl-2h-1,2,3,4-tetrazole, 5-2-bromophenyl-1h-tetraazole, 5-2-bromophenyl-1 h-tetrazole, 1h-tetrazole, 5-2-bromophenyl
Formula Weight 225.05
Percent Purity ≥98%
Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.