Learn More
5-(2-Aminoethyl)-2-methoxyphenol Hydrochloride (4-O-Methyldopamine Hydrochloride), Mikromol™
Product Code. 30050821
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050821 100 mg 100mg
30051038 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050821 Supplier Mikromol Supplier No. MM0384.02
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(2-Aminoethyl)-2-methoxyphenol Hydrochloride (4-O-Methyldopamine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(2-Aminoethyl)-2-methoxyphenol Hydrochloride (4-O-Methyldopamine Hydrochloride)
CAS 645-33-0
Analyte or Component Names 5-(2-Aminoethyl)-2-methoxyphenol Hydrochloride (4-O-Methyldopamine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C9 H13 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym Dopamine Hydrochloride Imp. A (EP), Dopamine Imp. A (EP), 5-(2-Aminoethyl)-2-methoxyphenol Hydrochloride, Dopamine Hydrochloride Imp. A (EP) as Hydrochloride, 4-O-Methyldopamine Hydrochloride, Dopamine Hydrochloride Impurity A as Hydrochloride, Dopamine Impurity A as Hydrochloride
InChI Formula InChI=1S/C9H13NO2.ClH/c1-12-9-3-2-7(4-5-10)6-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H
SMILES Cl.COc1ccc(CCN)cc1O
IUPAC Name 5-(2-aminoethyl)-2-methoxyphenol;hydrochloride
Molecular Weight (g/mol) 203.67
Quantity 100 mg
Formula Weight 203.0713 g/mol