missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium Chloride Hydrochloride (Acetylthiamine Hydrochloride), Mikromol™

Product Code. 17952286
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17952286

Brand: Mikromol MM0049.180025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium Chloride Hydrochloride (Acetylthiamine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamine Acetate Hydrochloride
CAS 1037-29-2
Analyte or Component Names Thiamine Acetate Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C14H20Cl2N4O2S
Certifications/Compliance ISO 9001
Synonym Thiazolium, 5-[2-(acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-, chloride, hydrochloride (1:1:1), Thiamine, acetate (ester), monohydrochloride (8CI), Thiamine, acetate, hydrochloride (7CI), Thiazolium, 5-[2-(acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-, chloride, monohydrochloride (9CI), Thiamine Hydrochloride Imp. G (EP) as Chloride Hydrochloride
InChI Formula InChI=1S/C14H19N4O2S.2ClH/c1-9-13(4-5-20-11(3)19)21-8-18(9)7-12-6-16-10(2)17-14(12)15;;/h6,8H,4-5,7H2,1-3H3,(H2,15,16,17);2*1H/q+1;;/p-1
SMILES Cl.[Cl-].CC(=O)OCCc1sc[n+](Cc2cnc(C)nc2N)c1C
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl acetate;chloride;hydrochloride
Molecular Weight (g/mol) 379.31
Quantity 25 mg
Formula Weight 378.0684 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.