Learn More
5,10,15-Triphenylcorrole, TRC
Specifications
Specifications
| Chemical Name or Material | 5,10,15-Triphenylcorrole |
| CAS | 246231-45-8 |
| Molecular Formula | C37H26N4 |
| Quantity | 50 mg |
| Synonym | 1,2,3,7,8,12,13,17,18,19-Decadehydro-21,22-dihydro-5,10,15-triphenylcorrin, meso-Triphenylcorrole |
| InChI Formula | InChI=1S/C37H26N4/c1-4-10-24(11-5-1)35-29-18-16-27(38-29)28-17-19-30(39-28)36(25-12-6-2-7-13-25)32-21-23-34(41-32)37(26-14-8-3-9-15-26)33-22-20-31(35)40-33/h1-23,38-40H/b28-27-,35-29-,35-31-,36-30-,36-32-,37-33-,37-34- |
| SMILES | N1\C\2=C(/C3=N\C(=C(\c4ccccc4)/c5ccc([nH]5)c6ccc([nH]6)\C(=C/1\C=C2)\c7ccccc7)\C=C3)\c8ccccc8 |
| IUPAC Name | (5Z,9Z,14Z)-5,10,15-triphenyl-22,24-dihydro-21H-corrin |
| Molecular Weight (g/mol) | 526.63 |
| Formula Weight | 526.2157 |