Learn More
(4S)-(-)-Isopropyl-2-oxazolidinone, 98%
Product Code. 11407916
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407916 1 g 1g
11417916 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11407916 Supplier Thermo Scientific Alfa Aesar Supplier No. A14029.03
Request Bulk or Custom Format
CAS: 17016-83-0 | C6H11NO2 | 129.159 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17016-83-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00010847
InChI Key YBUPWRYTXGAWJX-RXMQYKEDSA-N
Synonym s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
PubChem CID 7157133
IUPAC Name (4S)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1COC(=O)N1
Specifications
Melting Point 70°C to 74°C
Beilstein 3588198
Quantity 1 g
Formula Weight 129.16
Optical Rotation −17° (c=6 in Ethanol)
Percent Purity 98%
Chemical Name or Material (4S)-(-)-Isopropyl-2-oxazolidinone