Learn More
(4S)-6-Chloro-4-(2-cyclopropylethynyl)-3,4-dihydro-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)-quinazolinone, TRC
Specifications
Specifications
| Chemical Name or Material | (4S)-6-Chloro-4-(2-cyclopropylethynyl)-3,4-dihydro-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)-quinazolinone |
| CAS | 247565-04-4 |
| Molecular Formula | C22H18ClF3N2O |
| Quantity | 100 mg |
| Synonym | (4S)-6-Chloro-4-(cyclopropylethynyl)-3,4-dihydro-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)-quinazolinone |
| InChI Formula | InChI=1S/C22H18ClF3N2O/c1-14(16-5-3-2-4-6-16)28-20(29)27-19-10-9-17(23)13-18(19)21(28,22(24,25)26)12-11-15-7-8-15/h2-6,9-10,13-15H,7-8H2,1H3,(H,27,29)/t14-,21+/m1/s1 |
| SMILES | O=C1NC2=C(C=C(Cl)C=C2)[C@](C(F)(F)F)(C#CC3CC3)N1[C@@H](C4=CC=CC=C4)C |
| IUPAC Name | (S)-6-chloro-4-(cyclopropylethynyl)-3-((R)-1-phenylethyl)-4-(trifluoromethyl)-3,4-dihydroquinazolin-2(1H)-one |
| Molecular Weight (g/mol) | 418.84 |
| Formula Weight | 418.11 |