Learn More
(4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)quinazolinone, TRC
Specifications
Specifications
| Chemical Name or Material | (4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)quinazolinone |
| CAS | 247565-03-3 |
| Molecular Formula | C17H14ClF3N2O2 |
| Quantity | 100 mg |
| Synonym | 4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)-quinazolinone (ACI) |
| InChI Formula | InChI=1S/C17H14ClF3N2O2/c1-10(11-5-3-2-4-6-11)23-15(24)22-14-8-7-12(18)9-13(14)16(23,25)17(19,20)21/h2-10,25H,1H3,(H,22,24)/t10-,16+/m1/s1 |
| SMILES | O=C1NC2=C(C=C(Cl)C=C2)[C@](O)(C(F)(F)F)N1[C@@H](C3=CC=CC=C3)C |
| IUPAC Name | (S)-6-chloro-4-hydroxy-3-((R)-1-phenylethyl)-4-(trifluoromethyl)-3,4-dihydroquinazolin-2(1H)-one |
| Molecular Weight (g/mol) | 370.75 |
| Formula Weight | 370.07 |