missing translation for 'onlineSavingsMsg'
Learn More

4',6-Diamidino-2-Phenylindole Dihydrochloride MP Biomedicals

Product Code. 11431021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
50 mg
Unit Size:
1mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431021 50 mg 50mg
11401021 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431021 Supplier MP Biomedicals Supplier No. 0215757450
  • ONLINE EXCLUSIVE PRICE
    422.00 EUR
    401.10 EUR / 50mg
    Save 20.90 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
IUPAC Name dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

Quantity 50 mg
Formula Weight 350.2
Chemical Name or Material 4′, 6-Diamidino-2-Phenylindole Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.