Learn More
Skytec™ 4530 Disposable 3-Panel Fold Flat Respirator - FFP3
Click to view available options
:
20/Pk.
Description
- Premium disposable fold flat respirator for easy storage, donning and doffing
- Secure, metal-free harness system with color coded textile elastic for easy identification of FFP level
- Soft, flexible and adjustable nose bridge for increased fit factor and comfort
- Multilayer electrostatic filter media for high-performing filtration with low breathing resistance
- Full-width internal faceseal, made from soft touch PVC for high comfort
- Ergonomic design for a secure fit on a wide range of face shapes
- High flow exhalation valve for low breathing resistance, color-coded for easy identification of FFP level
- Contoured noseseal for high compatibility with other types of PPE
- All variants complaint to EN 149:2001+A1:2009. Unvalved variant compliant to EN 14683:2019 for use in healthcare
- Latex-free and low-linting for use in a wide range of environments
- UK designed, developed and manufactured.
Specifications
Specifications
| Assigned Protection Factor (APF) | 20 |
| Certifications/Compliance | CE III , UKCA , UK Medical Device Directive: 93/42/EEC and 2007/47/EC , EU Medical Device Regulation: 2017/745 , EN 149:2000 + A1:2009 FFP3 NR , EN 14683:2019 Type IIR |
| Color | White |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polypropylene |
| Product Type | Healthcare Particulate Respirator and Surgical Mask |
| Size | One Size |
| With Exhalation Valve | No |
| Show More |