Learn More
4-(Trifluoromethyl)benzenediazonium tetrafluoroborate, TRC
Product Code. 30455701
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455701 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455701 Supplier Toronto Research Chemicals Supplier No. TRCT791650250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Trifluoromethyl)benzenediazonium tetrafluoroborate
CAS 36407-40-6
Molecular Formula C7H4F3N2 . BF4
Quantity 250 mg
Synonym 4-(Trifluoromethyl)-','tetrafluoroborate(1-) Benzenediazonium, Borate(1-)','tetrafluoro-','4-(trifluoromethyl)benzenediazonium, 4-Trifluoromethylphenyldiazonium tetrafluoroborate
InChI Formula InChI=1S/C7H4F3N2.BF4/c8-7(9,10)5-1-3-6(12-11)4-2-5;2-1(3,4)5/h1-4H;/q+1;-1
SMILES [F-].FB(F)F.FC(F)(F)c1ccc(cc1)[N+2]#N
IUPAC Name 4-(trifluoromethyl)benzenediazonium;tetrafluoroborate
Molecular Weight (g/mol) 259.92
Formula Weight 260.0356