missing translation for 'onlineSavingsMsg'
Learn More

4-Toluidine-d7, TRC

Product Code. 30179624
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179624 250 mg 250mg
30179035 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179624 Supplier Toronto Research Chemicals Supplier No. TRCT536217250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68693-08-3
Molecular Formula C7 D7 H2 N
Molecular Weight (g/mol) 114.196
Synonym Benzen-2,3,5,6-d4-amine, 4-(methyl-d3)- (9CI, ACI), 4-(Methyl-d3)benzen-2,3,5,6-d4-amine (ACI), 4-Toluidine-d7, p-Toluidine-d7, 4-Methylaniline-D7
IUPAC Name 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)aniline
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)C([2H])([2H])[2H]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3/i1D3,2D,3D,4D,5D
Formula Weight 114.117
Chemical Name or Material 4-Toluidine-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.