missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Thien-3-ylphenyl)methylamine, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 876317-18-9 |
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Molecular Formula | C11H11NS |
| MDL Number | MFCD08271959 |
| Quantity | 1 g |
| Synonym | 4-thien-3-ylphenyl methylamine, 4-thiophen-3-yl phenyl methanamine, benzenemethanamine,4-3-thienyl, 1-4-thiophen-3-yl phenyl methanamine, 4-thien-3-yl benzylamine, 4-thiophen-3-yl benzylamine, 4-3-thienyl phenyl methylamine, 4-thiophen-3-ylphenyl methanamine, 4-thien-3-yl phenyl methylamine |
| InChI Key | QTXFTOUAVPVJMZ-UHFFFAOYSA-N |
| SMILES | C1=CC(=CC=C1CN)C2=CSC=C2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction