Learn More
4-tert-Butylphenyl glycidyl ether, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3101-60-8 |
| Molecular Formula | C13H18O2 |
| MDL Number | MFCD00005136 |
| Quantity | 5 g |
| SMILES | CC(C1=CC=C(OCC2CO2)C=C1)(C)C |
| Molecular Weight (g/mol) | 206.28 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-tert-Butylphenyl glycidyl ether |