missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzylamine, 98%

Product Code. p-7048161
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385659 5 g 5g
11395659 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385659 Supplier Thermo Scientific Alfa Aesar Supplier No. L20394.06
Add to basket
This item is not returnable. View return policy

CAS: 39895-55-1 | C11H17N | 163.264 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39895-55-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00040754
InChI Key MPWSRGAWRAYBJK-UHFFFAOYSA-N
Synonym 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine
PubChem CID 2735655
IUPAC Name (4-tert-butylphenyl)methanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CN

Specifications

Density 0.927
Boiling Point 124°C (16 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.521
Quantity 5 g
UN Number UN2735
Beilstein 2961797
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.