missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfophthalic acid, tech., 50% w/w aq. soln., Thermo Scientific™

Product Code. p-7023179
Change view
Click to view available options
Quantity:
2500 g
Unit Size:
2500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477787 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477787 Supplier Thermo Scientific Alfa Aesar Supplier No. A16318.0E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-08-7
Molecular Formula C8H6O7S
Molecular Weight (g/mol) 246.189
MDL Number MFCD00007494
InChI Key WNKQDGLSQUASME-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-sulfo, phthalic acid, 4-sulfo, 4-sulphophthalic acid, unii-t1sm5dom66, 4-sulfobenzene-1,2-dicarboxylic acid, t1sm5dom66, 4-sulfophthalic acid aq. soln., 5-sulfophthalic acid, acmc-209uex, 1, 4-sulfo
PubChem CID 6962
IUPAC Name 4-sulfophthalic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)C(=O)O

Specifications

Density 1.292
Refractive Index 1.447
Quantity 2500 g
UN Number UN3265
Formula Weight 246.2
Grade Technical
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material 4-Sulfophthalic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.