Learn More
4-Sulfamoylbenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Sulfamoylbenzoic Acid |
| CAS | 138-41-0 |
| Molecular Formula | C7H7NO4S |
| Quantity | 25 g |
| Synonym | 4-(Aminosulfonyl)benzoic Acid, 4-Carboxybenzenesulfonamide, 4-Sulfamoylbenzoic Acid, Benzoic Acid p-Sulfamide, Carzenide, Dirnate, NSC 22976, NSC 683540, p-Aminobenzoic Acid Sulfamide, p-Aminosulfonylbenzoic Acid, p-Carboxybenzenesulfonamide, p-Sulfamoylbenzoic Acid, p-Sulfamylbenzoic Acidp-Sulfonamidobenzoic Acid |
| InChI Formula | InChI=1S/C7H7NO4S/c8-13(11,12)6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)(H2,8,11,12) |
| SMILES | NS(=O)(=O)c1ccc(cc1)C(=O)O |
| IUPAC Name | 4-sulfamoylbenzoic acid |
| Molecular Weight (g/mol) | 201.2 |
| Formula Weight | 201.0096 |