Learn More
4-Pyridylethylmercaptan, Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Pyridylethylmercaptan Hydrochloride |
| CAS | 6298-11-9 |
| Molecular Formula | C7 H9 N S . Cl H |
| Quantity | 2.5 g |
| Synonym | 4-Pyridineethanethiol','hydrochloride (1:1), 4-Pyridineethanethiol','hydrochloride (6CI,8CI,9CI), 2-(Pyridin-4-yl)ethanethiol hydrochloride, 4-(2-Mercaptoethyl)pyridine hydrochloride |
| InChI Formula | InChI=1S/C7H9NS.ClH/c9-6-3-7-1-4-8-5-2-7;/h1-2,4-5,9H,3,6H2;1H |
| SMILES | Cl.SCCc1ccncc1 |
| IUPAC Name | 2-pyridin-4-ylethanethiol;hydrochloride |
| Molecular Weight (g/mol) | 175.68 |
| Formula Weight | 175.0222 |