Learn More
Alfa Aesar™ 4-Pyridineethanesulfonic acid, 99%
Product Code. 11362707
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362707 100 g 100g
11352707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11362707 Supplier Alfa Aesar™ Supplier No. L11906.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 53054-76-5
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00006452
InChI Key RGIIAYDCZSXHGL-UHFFFAOYSA-N
Synonym 4-pyridineethanesulfonic acid, 2-4-pyridyl ethanesulfonic acid, 4-pyridylethylsulfonic acid, 2-pyridin-4-yl ethanesulfonic acid, 4-pyridyl-2-ethyl sulfonic acid, 4-pyridineethanesulfonicacid, 2-4-pyridyl ethanesulphonic acid, 4-2-sulfoethyl pyridine, acmc-209l3n, 4-2-sulfoethyl-pyridine
PubChem CID 96722
IUPAC Name 2-pyridin-4-ylethanesulfonic acid
SMILES C1=CN=CC=C1CCS(=O)(=O)O
Specifications
Melting Point ∽295°C (decomposition)
Quantity 100 g
UN Number UN2585
Beilstein 136690
Formula Weight 187.22
Percent Purity 99%
Chemical Name or Material 4-Pyridineethanesulfonic acid