missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinecarboxylic Acid, Sigma-Aldrich™

Product Code. 15878380
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878380 250 g 250g
15868380 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15878380 Supplier Sigma Aldrich Supplier No. 8007360250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-22-1
Grade Research
Molecular Weight (g/mol) 123.11

Specifications

Melting Point 315°C to 319°C
Boiling Point 260°C/20 hPa
MDL Number MFCD00006429
Quantity 250 g
Synonym 4-Pyridinecarboxylic acid, iso-Nicotinic acid, 4-Pyridinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.