Learn More
4-Pyridinecarbaldehyde Oxime, Sigma-Aldrich™
Specifications
Specifications
| CAS | 696-54-8 |
| Melting Point | 130°C to 133°C |
| Molecular Formula | 4-(CH=NOH)C5H4N |
| MDL Number | MFCD00006446 |
| Quantity | 50 g |
| Synonym | 4-Pyridinecarbaldehyde oxime, Isonicotinaldoxime, 4-Pyridinecarbaldehyde oxime |
| Molecular Weight (g/mol) | 122.12 |
| Grade | Research |