missing translation for 'onlineSavingsMsg'
Learn More

4α-Phorbol-12-Myristate-13-acetate, 99%, MP Biomedicals™

Product Code. 11474252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474252 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474252 Supplier MP Biomedicals Supplier No. 0215898205
  • ONLINE EXCLUSIVE PRICE
    1125.00 EUR
    1068.90 EUR / 5mg
    Save 56.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Possible carcinogen

TRUSTED_SUSTAINABILITY

Specifications

CAS 63597-44-4
Color White
Assay Percent Range 99%
Molecular Formula C36H56O8
MDL Number MFCD00153860
Quantity 5 mg
Synonym 4alpha-phorbol 12-myristate 13-acetate, phorbol-12-myristate-13-acetate, pma, 4alpha-pma, 4alpha-tpa, .alpha.-tpa, 4.alpha.-tpa, 4alpha-phorbol-12-myristate-13-acetate, 4, a-pma, acetoxy-dihydroxy-hydroxymethyl-tetramethyl-oxo-? yl tetradecanoate
InChI Key PHEDXBVPIONUQT-LQLWEASQSA-N
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
IUPAC Name (1S,2S,6S,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol) 616.84
PubChem CID 122634
Formula Weight 616.8
Chemical Name or Material 4α-Phorbol-12-Myristate-13-acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.