missing translation for 'onlineSavingsMsg'
Learn More

4-Phenanthrol, TRC

Product Code. 30418543
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418543 25 mg 25mg
30417266 10 mg -
30415637 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418543 Supplier Toronto Research Chemicals Supplier No. TRCP29501525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7651-86-7
Molecular Formula C14 H10 O
Molecular Weight (g/mol) 194.23
Synonym 4-Phenanthrenol (9CI, ACI), 4-Phenanthrol (6CI, 7CI, 8CI), 4-Hydroxyphenanthrene, NSC 171284
IUPAC Name phenanthren-4-ol
SMILES Oc1cccc2ccc3ccccc3c12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H10O/c15-13-7-3-5-11-9-8-10-4-1-2-6-12(10)14(11)13/h1-9,15H
Formula Weight 194.0732
Chemical Name or Material 4-Hydroxy-phenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.