missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15477567
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15477567

Brand: Thermo Scientific Alfa Aesar H35736.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(Pentafluorothio)benzaldehyde is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401892-84-0
Molecular Formula C7H5F5OS
Molecular Weight (g/mol) 232.17
MDL Number MFCD03788518
InChI Key OTOYBKNIUCEPMV-UHFFFAOYSA-N
Synonym 4-pentafluorothio benzaldehyde, 4-pentafluorosulfanyl benzaldehyde, 4-pentafluoro-$l^ 6-sulfanyl benzaldehyde, 4-pentafluoro-??-sulfanyl benzaldehyde, pubchem8518, 4-pentafluorosulfanyl-benzaldehyde, 4-formylphenylsulphur pentafluoride, 4-pentafluorosulphanyl benzaldehyde, 4-formylphenyl pentafluorosulfur vi, 4-pentafluoro-lambda∼6∼-sulfanyl benzaldehyde
PubChem CID 2779207
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
SMILES FS(F)(F)(F)(F)C1=CC=C(C=O)C=C1

Specifications

Quantity 1 g
Sensitivity Air sensitive
Formula Weight 232.17
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.