accessibility menu, dialog, popup

UserName

CAS RN 401892-84-0

OSFFFFF

CAS RN 401892-84-0

IUPAC Name: 4-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 4-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 232.17
Molecular Formula: C7H5F5OS
InChi Key: OTOYBKNIUCEPMV-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results