Learn More
4-Oxo-4H-chromene-3-carbaldehyde, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 4-Oxo-4H-chromene-3-carbaldehyde |
| CAS | 17422-74-1 |
| Molecular Formula | C10H6O3 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C10H6O3/c11-5-7-6-13-9-4-2-1-3-8(9)10(7)12/h1-6H |
| SMILES | O=CC1=COc2ccccc2C1=O |
| IUPAC Name | 4-oxochromene-3-carbaldehyde |
| Molecular Weight (g/mol) | 174.15 |
| Formula Weight | 174.0317 |