Learn More
4-Nitrophthalimide, 98%
Product Code. p-7081872
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419912 5 g 5g
11429912 25 g 25g
11439912 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11419912 Supplier Thermo Scientific Alfa Aesar Supplier No. A12040.06
Add to basket
CAS: 89-40-7 | C8H4N2O4 | 192.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-40-7
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005884
InChI Key ANYWGXDASKQYAD-UHFFFAOYSA-N
Synonym 4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione
PubChem CID 6969
IUPAC Name 5-nitroisoindole-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O
Specifications
Melting Point 200°C to 204°C
Quantity 5 g
Beilstein 180224
Formula Weight 192.13
Percent Purity 98%
Chemical Name or Material 4-Nitrophthalimide