missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroindole, 97%, Thermo Scientific™

Product Code. 11357266
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357266 100 mg 100mg
11337266 500 mg 500mg
11347266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357266 Supplier Thermo Scientific Alfa Aesar Supplier No. L09177.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4769-97-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00010056
InChI Key LAVZKLJDKGRZJG-UHFFFAOYSA-N
Synonym 4-nitroindole, indole, 4-nitro, 1h-indole, 4-nitro, pubchem7440, 4-nitro-1 h-indole, #, maybridge1_006408, acmc-209ka5, ksc236o2l
PubChem CID 145766
IUPAC Name 4-nitro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-]

Specifications

Melting Point 202°C to 207°C
Quantity 100 mg
Beilstein 136022
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 4-Nitroindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.