Learn More
4-Nitroimidazole, Mikromol
Description
4-Nitroimidazole
Specifications
Specifications
| Chemical Name or Material | 4-Nitroimidazole |
| CAS | 3034-38-6 |
| Analyte or Component Names | 4-Nitroimidazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H3 N3 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Metronidazole Imp. B (EP), 5-Nitro-1H-imidazole, 4-Nitro-1H-imidazole, 4(or 5)-Nitroimidazole, 1H-4-Nitroimidazole, 1H-5-Nitroimidazole, 4(5)-Nitroimidazole, 4-Nitroimidazole, 5-Nitroimidazole, NSC 50359, Metronidazole Impurity B |
| InChI Formula | InChI=1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5) |
| SMILES | [O-][N+](=O)c1c[nH]cn1 |
| IUPAC Name | 4-nitro-1H-imidazole |
| Show More |